Assay Details
Target Gene Details
Entrez Gene ID: | 108100 |
Gene Name: | brain-specific angiogenesis inhibitor 1-associated protein 2 |
Gene Aliases: |
IRSp53, R75030 |
Location: |
Chr.11:119942869-120006782 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baiap2 | NM_001037754.3 | NP_001032843.1 | ||
NM_001037755.3 | NP_001032844.2 | |||
NM_130862.4 | NP_570932.2 | |||
AF390178.1 | AAK68153.1 | |||
AF390179.1 | AAK81838.1 | |||
AK049469.1 | BAC33764.1 | |||
AK145924.1 | BAE26755.1 | |||
AK158579.1 | ||||
AK160401.1 | BAE35770.1 | |||
AK170190.1 | ||||
BC006620.1 | AAH06620.1 | |||
BC016411.1 | AAH16411.1 |
More Information
Set Membership: |
Intragenic Non-exonic |