Assay Details
Target Gene Details
Entrez Gene ID: | 11421 |
Gene Name: | angiotensin I converting enzyme (peptidyl-dipeptidase A) 1 |
Gene Aliases: |
AW208573, CD143 |
Location: |
Chr.11:105967945-105989964 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ace | NM_001281819.1 | NP_001268748.1 | ||
NM_207624.5 | NP_997507.1 | |||
AK154632.1 | BAE32728.1 | |||
AK161020.1 | BAE36151.1 | |||
BC034367.1 | AAH34367.1 | |||
BC040404.1 | AAH40404.1 | |||
BC083109.1 | AAH83109.1 | |||
BC110362.1 | AAI10363.1 | |||
J04946.1 | AAA37147.1 |
Target Gene Details
Entrez Gene ID: | 100379610 |
Gene Name: | predicted gene 11651 |
Gene Aliases: |
OTTMUSG00000003114 |
Location: |
Chr.11:105965607-105985694 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm11651 | NR_104047.1 | |||
AA145017.1 | ||||
AK016887.1 | ||||
BU938668.1 | ||||
CF106408.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |