Assay Details
Target Gene Details
Entrez Gene ID: | 11302 |
Gene Name: | apoptosis-associated tyrosine kinase |
Gene Aliases: |
AATYK, aatyk1, mKIAA0641 |
Location: |
Chr.11:120007311-120047182 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aatk | NM_001198785.1 | NP_001185714.1 | ||
NM_001198787.1 | NP_001185716.1 | |||
NM_007377.4 | NP_031403.2 | |||
AB093253.2 | BAC41437.2 | |||
AB288871.1 | ||||
AF011908.1 | AAB71837.1 | |||
AK036705.1 | BAC29541.1 | |||
AK147239.1 | 1 | 397 | BAE27789.1 | |
AK155100.1 | BAE33045.1 | |||
AY236858.1 | AAO92350.1 | |||
AY236859.1 | AAO92351.1 | |||
BC080846.1 | AAH80846.1 |
More Information
Set Membership: |
Intragenic Non-exonic |