Assay Details
Target Gene Details
Entrez Gene ID: | 216151 |
Gene Name: | polymerase (RNA) mitochondrial (DNA directed) |
Gene Aliases: |
1110018N15Rik, 4932416K13 |
Location: |
Chr.10:79736125-79746589 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polrmt | NM_172551.3 | 8 | 1490 | NP_766139.2 |
AK030039.1 | 8 | 1494 | ||
AK042071.1 | 6 | 1047 | BAE20618.1 | |
AK047545.1 | 8 | 1483 | ||
AK053356.1 | 8 | 1481 | BAC35360.1 | |
AK085666.1 | 8 | 1483 | BAC39500.1 | |
AK128956.1 | 8 | 1482 | ||
AK154181.1 | 8 | 1481 | ||
AK154729.1 | 8 | 1472 | BAE32793.1 | |
AK213762.1 | ||||
BC110697.1 | 8 | 1466 | AAI10698.1 | |
BC117940.1 | 8 | 1483 |
More Information
Set Membership: |
Intragenic Exonic |