Assay Details
Target Gene Details
Entrez Gene ID: | 18114 |
Gene Name: | ribosomal RNA processing 1 homolog (S. cerevisiae) |
Gene Aliases: |
AL033305, AL033306, AU018604, NNP-1, Nnp1 |
Location: |
Chr.10:78400362-78413043 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrp1 | NM_010925.2 | NP_035055.2 | ||
AF312394.1 | AAG30293.1 | |||
AK077540.1 | ||||
BC003913.1 | ||||
BC022910.1 | ||||
BC065993.1 | AAH65993.1 | |||
BC065994.1 | AAH65994.1 | |||
U79773.1 | AAC53286.1 | |||
U79774.1 | AAB63261.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |