Assay Details
Target Gene Details
Entrez Gene ID: | 66420 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide E |
Gene Aliases: |
2410021N14Rik, 25kDa, AW208866, RPB5, XAP4 |
Location: |
Chr.10:80035953-80039659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2e | NM_025554.2 | NP_079830.2 | ||
AK010567.1 | ||||
AK015823.1 | BAE43245.1 | |||
AK088610.1 | ||||
AK132109.1 | BAE20985.1 | |||
AK144668.1 | BAE25998.1 | |||
AK145201.1 | BAE26292.1 | |||
BC026842.1 | AAH26842.1 | |||
BC037681.1 | AAH37681.1 | |||
BC045521.1 | AAH45521.1 |
More Information
Set Membership: |
Intragenic Non-exonic |