Assay Details
Target Gene Details
Entrez Gene ID: | 102634101 |
Gene Name: | predicted gene, 31763 |
Gene Aliases: |
- |
Location: |
Chr.10:67005075-67041326 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm31763 | NR_110511.1 | |||
AK079018.1 |
Target Gene Details
Entrez Gene ID: | 28193 |
Gene Name: | receptor accessory protein 3 |
Gene Aliases: |
D10Ucla1 |
Location: |
Chr.10:67009189-67096988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Reep3 | NM_001204915.1 | NP_001191844.1 | ||
NM_178606.5 | NP_848721.1 | |||
AK005026.1 | ||||
AK008058.1 | BAB25434.1 | |||
AK012403.1 | BAB28218.1 | |||
AK028868.1 | ||||
AK047728.1 | BAC33141.1 | |||
AK079649.1 | BAC37714.1 | |||
AK154559.1 | ||||
AY562231.1 | AAT70676.1 | |||
BC004607.1 | AAH04607.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |