Assay Details
Target Gene Details
Entrez Gene ID: | 108037 |
Gene Name: | serine hydroxymethyltransferase 2 (mitochondrial) |
Gene Aliases: |
2700043D08Rik, AA408223, AA986903 |
Location: |
Chr.10:127517123-127522444 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shmt2 | NM_001252316.1 | NP_001239245.1 | ||
NM_028230.4 | NP_082506.1 | |||
AK012355.1 | BAB28184.1 | |||
AK037339.1 | BAC29790.1 | |||
AK051207.1 | BAC34556.1 | |||
AK162421.1 | BAE36907.1 | |||
AK169192.1 | BAE40968.1 | |||
AK215786.1 | ||||
BC004825.1 | AAH04825.1 | |||
BC051396.1 | AAH51396.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |