Assay Details
Target Gene Details
Entrez Gene ID: | 19386 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
A430087B05Rik, AI256741, NUP358 |
Location: |
Chr.10:58446852-58494155 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ranbp2 | NM_011240.3 | 1 | 99 | NP_035370.2 |
AK041932.1 | BAC31101.1 | |||
AK145306.1 |
More Information
Set Membership: |
Intragenic Exonic |