Assay Details
Target Gene Details
Entrez Gene ID: | 73681 |
Gene Name: | tRNA methyltransferase 11 |
Gene Aliases: |
2410075D05Rik, 3110045I18Rik, AW213713 |
Location: |
Chr.10:30534225-30602147 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt11 | NM_028604.2 | 9 | 943 | NP_082880.2 |
AF532979.1 | 9 | 943 | AAQ10286.1 | |
AK010722.1 | 9 | 885 | BAB27142.1 | |
BC048703.1 | 7 | 618 | AAH48703.1 |
More Information
Set Membership: |
Intragenic Exonic |