Assay Details
Target Gene Details
Entrez Gene ID: | 73075 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 6 |
Gene Aliases: |
2900084F20Rik |
Location: |
Chr.10:41490439-41514295 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil6 | NM_028430.1 | NP_082706.1 | ||
AI463820.1 | ||||
AK013818.1 | BAB29003.1 | |||
BC147756.1 | ||||
BC147757.1 | ||||
BP772786.1 | ||||
CA466046.1 |
More Information
Set Membership: |
Intragenic Non-exonic |