Assay Details
Target Gene Details
Entrez Gene ID: | 73132 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, Graves disease autoantigen), member 16 |
Gene Aliases: |
3110021G18Rik, GDA, GDC, HGT.1, ML7 |
Location: |
Chr.10:62920633-62946499 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a16 | NM_175194.2 | 1 | 123 | NP_780403.1 |
AK030865.1 | 1 | 123 | BAC27163.1 | |
BC062168.1 | 1 | 89 | AAH62168.1 | |
BC138983.1 | ||||
BC138984.1 |
More Information
Set Membership: |
Intragenic Exonic |