Assay Details
Target Gene Details
Entrez Gene ID: | 67569 |
Gene Name: | MGAT4 family, member C |
Gene Aliases: |
9130411I17Rik, GntIVh |
Location: |
Chr.10:101681487-102391851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4c | NM_001162368.1 | NP_001155840.1 | ||
NM_001162369.1 | NP_001155841.1 | |||
NM_001205098.1 | NP_001192027.1 | |||
NM_026243.4 | NP_080519.2 | |||
AK018574.1 | BAB31286.1 | |||
AK018672.1 | BAB31336.1 | |||
AK032009.1 | ||||
AK033482.1 | BAC28312.1 | |||
BB611341.1 | ||||
BC046987.1 | AAH46987.1 | |||
CD774691.1 |
More Information
Set Membership: |
Intragenic Non-exonic |