Assay Details
Target Gene Details
Entrez Gene ID: | 26408 |
Gene Name: | mitogen-activated protein kinase kinase kinase 5 |
Gene Aliases: |
7420452D20Rik, ASK, ASK1, MAPKKK5, Mekk5 |
Location: |
Chr.10:19934513-20142750 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map3k5 | NM_008580.4 | 9 | 1508 | NP_032606.4 |
AB006787.3 | 9 | 1560 | BAA23648.3 | |
BC116627.1 | 9 | 1508 | ||
BC116628.1 | 8 | 1484 | ||
BC133697.1 | 9 | 1508 |
More Information
Set Membership: |
Intragenic Exonic |