Assay Details
Target Gene Details
Entrez Gene ID: | 69736 |
Gene Name: | nucleoporin 37 |
Gene Aliases: |
2410003L22Rik, 2810039M17Rik |
Location: |
Chr.10:88146976-88178395 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nup37 | NM_027191.2 | NP_081467.2 | ||
NM_028334.4 | NP_082610.1 | |||
AK010370.1 | BAB26888.1 | |||
AK012890.1 | BAB28538.1 | |||
AK040463.1 | BAC30597.1 | |||
AK165537.1 | BAE38243.1 | |||
BC011102.1 | AAH11102.1 | |||
BC032180.1 | AAH32180.1 |
More Information
Set Membership: |
Intragenic Non-exonic |