Assay Details
Target Gene Details
Entrez Gene ID: | 69736 |
Gene Name: | nucleoporin 37 |
Gene Aliases: |
2410003L22Rik, 2810039M17Rik |
Location: |
Chr.10:88146976-88178395 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nup37 | NM_027191.2 | 3 | 366 | NP_081467.2 |
NM_028334.4 | 3 | 384 | NP_082610.1 | |
AK010370.1 | 3 | 310 | BAB26888.1 | |
AK012890.1 | 3 | 380 | BAB28538.1 | |
AK040463.1 | 3 | 367 | BAC30597.1 | |
AK165537.1 | 3 | 330 | BAE38243.1 | |
BC011102.1 | 3 | 350 | AAH11102.1 | |
BC032180.1 | 2 | 294 | AAH32180.1 | |
CJ072834.1 |
More Information
Set Membership: |
Intragenic Exonic |