Assay Details
Target Gene Details
Entrez Gene ID: | 26408 |
Gene Name: | mitogen-activated protein kinase kinase kinase 5 |
Gene Aliases: |
7420452D20Rik, ASK, ASK1, MAPKKK5, Mekk5 |
Location: |
Chr.10:19934513-20142750 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map3k5 | NM_008580.4 | 26 | 3693 | NP_032606.4 |
AB006787.3 | 26 | 3745 | BAA23648.3 | |
AK033726.1 | 2 | 125 | BAC28449.1 | |
AK078668.1 | 12 | 1489 | ||
BC116627.1 | 26 | 3693 | ||
BC116628.1 | 25 | 3669 | ||
BC133697.1 | 26 | 3693 |
More Information
Set Membership: |
Intragenic Exonic |