Assay Details
Target Gene Details
Entrez Gene ID: | 53605 |
Gene Name: | nucleosome assembly protein 1-like 1 |
Gene Aliases: |
AA407126, AI256722, D10Ertd68e, NAP-1 |
Location: |
Chr.10:111473192-111498150 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nap1l1 | NM_001146707.1 | NP_001140179.1 | ||
NM_015781.4 | NP_056596.1 | |||
AK004633.1 | ||||
AK007322.1 | BAE43200.1 | |||
AK012250.1 | BAB28118.1 | |||
AK032896.1 | BAC28075.1 | |||
AK050375.1 | BAC34219.1 | |||
AK136161.1 | BAE22850.1 | |||
AK145766.1 | BAE26637.1 | |||
AK152011.1 | ||||
AK153531.1 | ||||
AK169301.1 | BAE41057.1 | |||
AK195357.1 | ||||
AK216281.1 | ||||
BC076591.1 | AAH76591.1 | |||
D12618.1 | BAA02142.1 | |||
X61449.1 | CAA43689.1 |
More Information
Set Membership: |
Intragenic Non-exonic |