Assay Details
Target Gene Details
Entrez Gene ID: | 16000 |
Gene Name: | insulin-like growth factor 1 |
Gene Aliases: |
C730016P09Rik, Igf-1, Igf-I |
Location: |
Chr.10:87858270-87937047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf1 | NM_001111274.1 | NP_001104744.1 | ||
NM_001111275.1 | NP_001104745.1 | |||
NM_001111276.1 | NP_001104746.1 | |||
NM_010512.4 | NP_034642.2 | |||
NM_184052.3 | NP_908941.1 | |||
AF440694.1 | AAL34535.1 | |||
AK038119.1 | BAC29934.1 | |||
AK081019.1 | BAC38117.1 | |||
AK155435.1 | BAE33264.1 | |||
AK165471.1 | BAE38208.1 | |||
AY878192.1 | AAX61179.1 | |||
AY878193.1 | AAX61180.1 | |||
BC012409.1 | AAH12409.1 | |||
CT010364.1 | CAJ18571.1 | |||
X04480.1 | CAA28168.1 | |||
X04482.1 | CAA28170.1 |
More Information
Set Membership: |
Intragenic Non-exonic |