Assay Details
Target Gene Details
Entrez Gene ID: | 70208 |
Gene Name: | mediator complex subunit 23 |
Gene Aliases: |
130kDa, 3000002A17Rik, Crsp3, ESTM7, Sur2, X83317, mKIAA1216 |
Location: |
Chr.10:24869986-24913529 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med23 | NM_001166416.1 | NP_001159888.1 | ||
NM_027347.3 | NP_081623.3 | |||
AF507918.1 | AAM28897.1 | |||
AK028545.1 | BAC26001.1 | |||
AK042346.1 | ||||
AK129312.1 | BAC98122.1 | |||
BC050916.1 | AAH50916.1 | |||
CJ100894.1 |
More Information
Set Membership: |
Intragenic Non-exonic |