Assay Details
Target Gene Details
Entrez Gene ID: | 103199 |
Gene Name: | FIG4 homolog (S. cerevisiae) |
Gene Aliases: |
A530089I17Rik, AI326867, Sac3 |
Location: |
Chr.10:41188172-41303241 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fig4 | NM_133999.1 | NP_598760.1 | ||
AK041196.1 | ||||
AK169881.1 | BAE41433.1 | |||
AK172130.1 | BAE42839.1 | |||
AK172341.1 | ||||
AK172926.1 | BAD32204.1 | |||
AK181110.1 | ||||
AK204518.1 | ||||
BC015295.1 | AAH15295.1 | |||
BC031887.1 | AAH31887.1 |
More Information
Set Membership: |
Intragenic Non-exonic |