Assay Details
Target Gene Details
Entrez Gene ID: | 327762 |
Gene Name: | DNA replication helicase 2 homolog (yeast) |
Gene Aliases: |
Dna2l, E130315B21Rik |
Location: |
Chr.10:62946980-62974188 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dna2 | NM_177372.3 | 6 | 860 | NP_796346.2 |
AK028381.1 | 6 | 837 | BAC25919.1 | |
AK129051.1 | 6 | 845 | BAC97861.1 | |
BC025182.1 | AAH25182.1 | |||
BC030382.1 | 6 | 802 | AAH30382.1 | |
BC115716.1 | 6 | 814 | ||
CO433698.1 | 4 | 431 |