Assay Details
Target Gene Details
Entrez Gene ID: | 237362 |
Gene Name: | neuropeptide FF receptor 1 |
Gene Aliases: |
Gm236, Gpr147, NPFF1, OT7T022 |
Location: |
Chr.10:61595484-61628762 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npffr1 | NM_001177511.1 | 2 | 380 | NP_001170982.1 |
AY255578.1 | AAO85090.1 |
More Information
Set Membership: |
Intragenic Exonic |