Assay Details
Target Gene Details
Entrez Gene ID: | 56422 |
Gene Name: | Hbs1-like (S. cerevisiae) |
Gene Aliases: |
2810035F15Rik, AI326327, eRFS |
Location: |
Chr.10:21295979-21368889 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hbs1l | NM_001042593.1 | NP_001036058.1 | ||
NM_019702.2 | NP_062676.2 | |||
AF087672.1 | AAD23351.1 | |||
AK146678.1 | BAE27353.1 | |||
AK166140.1 | BAE38593.1 | |||
AK168671.1 | BAE40521.1 | |||
AK169281.1 | BAE41039.1 | |||
AK173091.1 | BAD32369.1 | |||
BC010251.1 | AAH10251.1 |
More Information
Set Membership: |
Intragenic Non-exonic |