Assay Details
Target Gene Details
Entrez Gene ID: | 16414 |
Gene Name: | integrin beta 2 |
Gene Aliases: |
2E6, AI528527, Cd18, LAD, LCAMB, Lfa1, MF17 |
Location: |
Chr.10:77530328-77565681 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb2 | NM_008404.4 | NP_032430.2 | ||
AK088435.1 | BAC40350.1 | |||
AK136502.1 | BAE23013.1 | |||
AK161236.1 | ||||
AK169587.1 | ||||
BC145644.1 | ||||
M31039.1 | AAA39325.1 | |||
X14951.1 | CAA33077.1 |
More Information
Set Membership: |
Intragenic Non-exonic |