Assay Details
Target Gene Details
Entrez Gene ID: | 23943 |
Gene Name: | extended synaptotagmin-like protein 1 |
Gene Aliases: |
Fam62a, Mbc2, vp115 |
Location: |
Chr.10:128510248-128525859 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esyt1 | NM_011843.2 | 18 | 2029 | NP_035973.1 |
AF098634.1 | 9 | 822 | AAD10190.1 | |
AK044656.1 | 18 | 2039 | BAC32020.1 | |
AK054147.1 | 9 | 813 | ||
AK080094.1 | 18 | 2042 | ||
AK088421.1 | 18 | 2043 | ||
AK142877.1 | 1 | 1518 | ||
BC011482.1 | 18 | 2029 | AAH11482.1 |