Assay Details
Target Gene Details
Entrez Gene ID: | 15353 |
Gene Name: | high mobility group 20B |
Gene Aliases: |
AW610687, BRAF35, Hmgx2, Hmgxb2, Smarce1r |
Location: |
Chr.10:81346046-81350511 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hmg20b | NM_001163165.1 | 4 | 464 | NP_001156637.1 |
NM_001163166.1 | 4 | 361 | NP_001156638.1 | |
NM_010440.3 | 3 | 548 | NP_034570.1 | |
AF146224.1 | 3 | 531 | AAF66708.1 | |
AK153816.1 | 4 | 362 | BAE32194.1 | |
AK155615.1 | 4 | 465 | BAE33346.1 | |
AK155652.1 | 4 | 366 | BAE33368.1 | |
AK155890.1 | 3 | 424 | BAE33486.1 | |
AK187890.1 | ||||
AL355734.1 | 3 | 543 | CAB90813.1 | |
BC003924.1 | 3 | 436 | ||
BC011334.1 | 4 | 397 | AAH11334.1 |