Assay Details
Target Gene Details
Entrez Gene ID: | 16404 |
Gene Name: | integrin alpha 7 |
Gene Aliases: |
[a]7, alpha7 |
Location: |
Chr.10:128933775-128958286 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga7 | NM_008398.2 | 12 | 1835 | NP_032424.2 |
AK157612.1 | 12 | 1842 | BAE34134.1 | |
L23423.1 | 12 | 1833 | AAA16600.1 |
More Information
Set Membership: |
Intragenic Exonic |