Assay Details
Target Gene Details
Entrez Gene ID: | 216190 |
Gene Name: | adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2 |
Gene Aliases: |
Dip3b |
Location: |
Chr.10:83600033-83648854 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Appl2 | NM_145220.2 | 15 | 1410 | NP_660255.1 |
AK160075.1 | 15 | 1389 | BAE35609.1 | |
AK160105.1 | 15 | 1386 | BAE35632.1 | |
AK161003.1 | 12 | 2962 | ||
AK168403.1 | 15 | 1373 | ||
AK168748.1 | 15 | 1366 | BAE40588.1 | |
AK171512.1 | 15 | 1366 | BAE42499.1 | |
AK195688.1 | ||||
AK198914.1 | ||||
AY113706.1 | 15 | 1412 | AAM55532.1 | |
BC002232.1 | 3 | 221 | AAH02232.1 | |
BC048906.1 | 15 | 1348 | AAH48906.1 |
More Information
Set Membership: |
Intragenic Exonic |