Assay Details
Target Gene Details
Entrez Gene ID: | 108707 |
Gene Name: | family with sequence similarity 207, member A |
Gene Aliases: |
1810008A18Rik, AI303072 |
Location: |
Chr.10:77486655-77537969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam207a | NM_133998.3 | NP_598759.1 | ||
AF391115.1 | AAL34506.1 | |||
AK076516.1 | BAC36374.1 | |||
AK088306.1 | BAC40272.1 | |||
AK160157.1 | BAE35663.1 | |||
BB840302.1 | ||||
BC034216.1 | AAH34216.1 |