Assay Details
Target Gene Details
Entrez Gene ID: | 74048 |
Gene Name: | RIKEN cDNA 4632428N05 gene |
Gene Aliases: |
Dies1, PD-1H, VISTA |
Location: |
Chr.10:60346851-60428381 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4632428N05Rik |
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | 28 | 3703 | NP_001239564.1 |
NM_023370.3 | 29 | 3709 | NP_075859.2 | |
NR_045556.1 | 28 | 3703 | ||
AF308939.1 | 27 | 3302 | AAG52817.1 | |
AJ888000.1 | 26 | 3296 | CAI59609.1 | |
AJ888001.1 | 26 | 3296 | CAI59610.1 | |
AJ888002.1 | 26 | 3296 | CAI59611.1 | |
AY026062.1 | 27 | 3311 | AAK07670.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |