Assay Details
Target Gene Details
Entrez Gene ID: | 104248 |
Gene Name: | calcineurin binding protein 1 |
Gene Aliases: |
A330070M20Rik, Cain, Ppp3in |
Location: |
Chr.10:75646110-75764358 on Build GRCm38 |
Assay Gene Location: | Within Exon 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabin1 | NM_172549.3 | 35 | 6532 | NP_766137.2 |
AK039606.1 | 9 | 2235 | ||
AK078480.1 | 7 | 1451 | BAC37299.1 | |
AK169961.1 | 7 | 1389 | BAE41484.1 | |
AK171567.1 | 7 | 1417 | ||
BC051668.1 | 2 | 164 | AAH51668.1 | |
BC056651.1 | 4 | 739 | AAH56651.1 | |
BC057551.1 | AAH57551.1 | |||
BC150825.1 | 34 | 6096 |
More Information
Set Membership: |
Intragenic Exonic |