Assay Details
Target Gene Details
Entrez Gene ID: | 237320 |
Gene Name: | aldehyde dehydrogenase 8 family, member A1 |
Gene Aliases: |
Raldh4 |
Location: |
Chr.10:21377300-21396578 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh8a1 | NM_178713.4 | NP_848828.1 | ||
AF510322.1 | AAO64246.1 | |||
AK050298.1 | BAC34173.1 | |||
AK143752.1 | BAE25524.1 | |||
AK165447.1 | ||||
BC038493.1 | AAH38493.1 |
More Information
Set Membership: |
Intragenic Non-exonic |