Assay Details
Target Gene Details
Entrez Gene ID: | 72068 |
Gene Name: | CCR4-NOT transcription complex, subunit 2 |
Gene Aliases: |
2600016M12Rik, 2810470K03Rik, AA537049, AA959607, AW557563, C79650 |
Location: |
Chr.10:116485161-116581535 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnot2 | NM_001037846.3 | NP_001032935.2 | ||
NM_001037847.2 | NP_001032936.2 | |||
NM_028082.2 | NP_082358.2 | |||
NR_024327.1 | ||||
AK011231.1 | BAB27481.1 | |||
AK078121.1 | BAC37134.1 | |||
AK149767.1 | BAE29072.1 | |||
AK167834.1 | BAE39856.1 | |||
BC043133.1 | AAH43133.1 | |||
BC063105.1 | AAH63105.1 | |||
BC065171.1 | AAH65171.1 | |||
BC090624.1 | AAH90624.1 |
More Information
Set Membership: |
Intragenic Non-exonic |