Assay Details
Target Gene Details
Entrez Gene ID: | 57279 |
Gene Name: | solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20 |
Gene Aliases: |
1110007P09Rik, C78826, Cact, mCAC |
Location: |
Chr.9:108662098-108684641 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a20 | NM_020520.4 | NP_065266.1 | ||
AK075624.1 | BAC35865.1 | |||
AK152436.1 | BAE31218.1 | |||
AK161086.1 | 1 | 213 | ||
BC029733.1 | AAH29733.1 | |||
BC052871.1 | AAH52871.1 |
More Information
Set Membership: |
Intragenic Non-exonic |