Assay Details
Target Gene Details
Entrez Gene ID: | 66163 |
Gene Name: | mitochondrial ribosomal protein L4 |
Gene Aliases: |
1110017G11Rik |
Location: |
Chr.9:21002605-21008839 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl4 | NM_023167.2 | NP_075656.2 | ||
AB041599.1 | BAA95082.1 | |||
AB049634.1 | BAB40839.1 | |||
AK002464.1 | BAB22118.1 | |||
AK003746.2 | ||||
AK216490.1 | ||||
BC037064.1 | AAH37064.1 | |||
BC039983.1 | AAH39983.2 | |||
BC061095.1 | AAH61095.1 |
More Information
Set Membership: |
Intragenic Non-exonic |