Assay Details
Target Gene Details
Entrez Gene ID: | 53356 |
Gene Name: | eukaryotic translation initiation factor 3, subunit G |
Gene Aliases: |
44kDa, D0Jmb4, Eif3s4, TU-189B2, eIF3-p44, p44 |
Location: |
Chr.9:20894349-20898590 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3g | NM_016876.3 | NP_058572.2 | ||
AF108214.1 | AAF14221.1 | |||
AK038249.1 | BAC29952.1 | |||
AK168362.1 | BAE40297.1 | |||
AK214405.1 | ||||
BC008511.1 | AAH08511.1 | |||
M88758.1 |
More Information
Set Membership: |
Intragenic Non-exonic |