Assay Details
Target Gene Details
Entrez Gene ID: | 30959 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 25 |
Gene Aliases: |
AW047046, GRTH |
Location: |
Chr.9:35541848-35558470 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx25 | NM_013932.4 | 10 | 1231 | NP_038960.2 |
AF142630.3 | 10 | 1162 | AAF21361.2 | |
AK050693.1 | 10 | 1226 | BAC34384.1 | |
AK078340.1 | 10 | 1190 | BAC37227.1 | |
BC024852.1 | 10 | 1175 | AAH24852.1 | |
BC061130.1 | 10 | 1186 |