Assay Details
Target Gene Details
Entrez Gene ID: | 110446 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 1 |
Gene Aliases: |
6330585C21Rik, Acat |
Location: |
Chr.9:53580522-53610350 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acat1 | NM_144784.3 | 6 | 570 | NP_659033.1 |
AJ510150.1 | 6 | 562 | CAD52869.1 | |
AK032097.1 | 6 | 541 | BAC27697.1 | |
AK075593.1 | 6 | 542 | ||
AK081715.1 | 6 | 570 | BAC38304.1 | |
AK146241.1 | 6 | 525 | ||
AK163402.1 | 6 | 574 | BAE37335.1 | |
AK169771.1 | 6 | 563 | BAE41356.1 | |
BC024763.1 | 6 | 542 | AAH24763.1 |
More Information
Set Membership: |
Intragenic Exonic |