Assay Details
Target Gene Details
Entrez Gene ID: | 235040 |
Gene Name: | autophagy related 4D, cysteine peptidase |
Gene Aliases: |
9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4 |
Location: |
Chr.9:21265285-21274837 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atg4d | NM_153583.10 | NP_705811.8 | ||
AK036282.1 | ||||
AK036571.1 | BAC29484.1 | |||
AK081002.1 | BAC38110.1 | |||
AK081605.1 | BAC38269.1 | |||
AK087258.1 | ||||
AK154130.1 | BAE32395.1 | |||
BB857077.1 | ||||
BC030861.1 | AAH30861.1 | |||
BC069851.1 | AAH69851.1 |
More Information
Set Membership: |
Intragenic Non-exonic |