Assay Details
Target Gene Details
Entrez Gene ID: | 19656 |
Gene Name: | RNA binding motif protein, X linked-like-1 |
Gene Aliases: |
Hnrpg, Rbmx, Rbmxrt |
Location: |
Chr.8:78505269-78508928 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbmxl1 | NM_001252089.1 | NP_001239018.1 | ||
NM_009033.2 | NP_033059.2 | |||
AF031568.1 | AAB86639.1 | |||
AK041927.1 | BAC31099.1 | |||
AK088213.1 | ||||
AK149658.1 | ||||
AK149764.1 | ||||
AK177134.1 | ||||
AK180901.1 | ||||
BC011441.1 | AAH11441.1 | |||
BC089350.1 | AAH89350.1 | |||
BY262810.1 |