Assay Details
Target Gene Details
Entrez Gene ID: | 212528 |
Gene Name: | tRNA methyltransferase 1 |
Gene Aliases: |
6720406L13Rik, 6720477L24, D8Ertd812e |
Location: |
Chr.8:84688725-84699808 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt1 | NM_001164559.1 | NP_001158031.1 | ||
NM_001164560.1 | NP_001158032.1 | |||
NM_198020.2 | NP_932137.2 | |||
AK032945.1 | ||||
AK040715.1 | 3 | 676 | ||
AK078395.1 | ||||
AK084775.1 | ||||
AK139463.1 | ||||
AK155251.1 | ||||
AK159467.1 | BAE35108.1 | |||
AK170412.1 | BAE41778.1 | |||
AK171002.1 | BAE42173.1 | |||
AK177323.1 | ||||
AK183397.1 | ||||
AK188154.1 | ||||
AK207177.1 | ||||
AK210059.1 | ||||
AK216489.1 | ||||
BC027404.1 | AAH27404.1 |
More Information
Set Membership: |
Intragenic Non-exonic |