Assay Details
Target Gene Details
Entrez Gene ID: | 66432 |
Gene Name: | solute carrier family 7, member 6 opposite strand |
Gene Aliases: |
2010007L18Rik, 2400002F02Rik |
Location: |
Chr.8:106200438-106210992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a6os | NM_001007567.2 | NP_001007568.1 | ||
AK010254.1 | 4 | 756 | ||
AK146733.1 | BAE27395.1 | |||
AK171806.1 | 2 | 2476 | ||
BC054118.1 | AAH54118.1 |