Assay Details
Target Gene Details
Entrez Gene ID: | 22227 |
Gene Name: | uncoupling protein 1 (mitochondrial, proton carrier) |
Gene Aliases: |
AI385626, Slc25a7, Ucp |
Location: |
Chr.8:83290348-83298456 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ucp1 | NM_009463.3 | NP_033489.1 | ||
AK002759.1 | ||||
AK076454.1 | ||||
AV248262.2 | ||||
BC012701.1 | AAH12701.1 | |||
U63419.1 | AAB05870.1 |
More Information
Set Membership: |
Intragenic Non-exonic |