Assay Details
Target Gene Details
Entrez Gene ID: | 26386 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
HSF 4, HSTF 4, HSTF4, ldis1, mHSF4 |
Location: |
Chr.8:105269748-105275845 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsf4 | NM_001256042.1 | NP_001242971.1 | ||
NM_001256044.1 | NP_001242973.1 | |||
NM_011939.3 | NP_036069.1 | |||
NR_045689.1 | ||||
AB029349.1 | BAA84583.1 | |||
AB029350.1 | BAA84584.1 | |||
AF160966.1 | AAF80399.1 | |||
AK148056.1 | BAE28316.1 | |||
BC138130.1 |
More Information
Set Membership: |
Intragenic Non-exonic |