Assay Details
Target Gene Details
Entrez Gene ID: | 57312 |
Gene Name: | mitochondrial ribosomal protein S31 |
Gene Aliases: |
1500002D03Rik, AA960481, Imogen38, Imogen44 |
Location: |
Chr.8:22410472-22431044 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrps31 | NM_020560.2 | NP_065585.1 | ||
AK005110.1 | BAB23825.1 | |||
AK011825.1 | ||||
AK019202.1 | ||||
AK078073.1 | ||||
AK161913.1 | BAE36630.1 | |||
BC018475.1 | AAH18475.1 | |||
BC027430.1 | AAH27430.1 | |||
Z46966.1 | CAA87087.1 |
More Information
Set Membership: |
Intragenic Non-exonic |