Assay Details
Target Gene Details
Entrez Gene ID: | 26386 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
HSF 4, HSTF 4, HSTF4, ldis1, mHSF4 |
Location: |
Chr.8:105269748-105275845 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsf4 | NM_001256042.1 | 10 | 1211 | NP_001242971.1 |
NM_001256044.1 | 9 | 1102 | NP_001242973.1 | |
NM_011939.3 | 10 | 1121 | NP_036069.1 | |
NR_045689.1 | 10 | 1107 | ||
AB029349.1 | 10 | 1143 | BAA84583.1 | |
AB029350.1 | 10 | 1048 | BAA84584.1 | |
AF160966.1 | 10 | 1138 | AAF80399.1 | |
AK148056.1 | 9 | 1074 | BAE28316.1 | |
AK177833.1 | ||||
AK184314.1 | ||||
AK215529.1 | ||||
BC138130.1 | 10 | 1211 |
More Information
Set Membership: |
Intragenic Exonic |