Assay Details
Target Gene Details
Entrez Gene ID: | 67873 |
Gene Name: | methylthioribose-1-phosphate isomerase homolog (S. cerevisiae) |
Gene Aliases: |
2410018C20Rik |
Location: |
Chr.8:84249629-84257337 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mri1 | NM_026423.4 | 4 | 825 | NP_080699.3 |
AK007709.1 | 4 | 786 | ||
AK010538.1 | 4 | 797 | BAB27014.1 | |
AK013882.1 | 4 | 794 | BAB29034.1 | |
AK047287.1 | 4 | 828 | BAC33016.1 | |
AK144807.1 | 4 | 768 | ||
AK164894.1 | 4 | 794 | BAE37957.1 | |
AK177142.1 | 1 | 49 | ||
AK182796.1 | 1 | 50 | ||
AK185486.1 | 2 | 134 | ||
BC025049.1 | 4 | 792 | AAH25049.1 |
More Information
Set Membership: |
Intragenic Exonic |