Assay Details
Target Gene Details
Entrez Gene ID: | 192156 |
Gene Name: | mevalonate (diphospho) decarboxylase |
Gene Aliases: |
C78718, Mpd |
Location: |
Chr.8:122433596-122443422 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mvd | NM_138656.2 | NP_619597.2 | ||
NR_028354.1 | ||||
AJ309922.1 | CAC35731.1 | |||
AK089354.1 | BAC40852.1 | |||
AK134803.1 | BAE22291.1 | |||
AK153439.1 | ||||
AK154883.1 | BAE32901.1 | |||
AK170007.1 | ||||
AK170773.1 | BAE42019.1 | |||
BC008526.1 | AAH08526.1 |
More Information
Set Membership: |
Intragenic Non-exonic |