Assay Details
Target Gene Details
Entrez Gene ID: | 20312 |
Gene Name: | chemokine (C-X3-C motif) ligand 1 |
Gene Aliases: |
AB030188, ABCD-3, AI848747, CX3C, Cxc3, D8Bwg0439e, Scyd1 |
Location: |
Chr.8:94772180-94782427 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cx3cl1 | NM_009142.3 | 2 | 217 | NP_033168.2 |
AF010586.1 | 2 | 171 | AAB66331.1 | |
AF071549.1 | 2 | 208 | AAD41620.1 | |
AK040220.1 | 2 | 162 | BAC30544.1 | |
AK078285.1 | 2 | 194 | ||
AK138214.1 | 2 | 194 | BAE23585.1 | |
AK138472.1 | 2 | 194 | BAE23673.1 | |
AK138574.1 | 2 | 194 | BAE23702.1 | |
AK154646.1 | 2 | 194 | BAE32737.1 | |
BC006650.1 | 2 | 171 | AAH06650.1 | |
BC054838.1 | 2 | 177 | AAH54838.1 | |
CB526167.1 | 2 | 217 | ||
U92565.1 | 2 | 132 | AAB71763.1 |
More Information
Set Membership: |
Intragenic Exonic |